Assay Details
Gene Symbol: CCDC157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103333.2 | NP_001096803.2 | 9 - 10 | 1876 | 103 | |
GenBank mRNA | BC134591.1 | - | 9 - 10 | 1878 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
