Assay Details
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103341.1 | NP_001096811.1 | 5 - 6 | 701 | 117 | |
GenBank mRNA | BC142481.1 | - | 5 - 6 | 701 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
