Assay Details
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105019.1 | NP_001098489.1 | 1 - 2 | 95 | 74 | |
GenBank mRNA | BC142236.1 | - | 1 - 2 | 95 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC142236.1, NM_001105019.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
