Assay Details
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105019.1 | NP_001098489.1 | 6 - 7 | 638 | 68 | |
GenBank mRNA | BC142236.1 | - | 6 - 7 | 638 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
