Assay Details
Gene Symbol: CRTAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105457.1 | NP_001098927.1 | 6 - 7 | 783 | 78 | |
GenBank mRNA | BC134770.1 | - | 6 - 7 | 783 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
