Assay Details
Gene Symbol: FLACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105461.1 | NP_001098931.1 | 2 - 3 | 525 | 138 | |
GenBank mRNA | BC123837.1 | - | 2 - 3 | 525 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
