Assay Details
Gene Symbol: PCYOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105474.2 | NP_001098944.1 | 5 - 6 | 897 | 113 | |
GenBank mRNA | BC149440.1 | - | 5 - 6 | 898 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
