Assay Details
Gene Symbol: FAM228B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105495.2 | NP_001098965.1 | 6 - 7 | 738 | 128 | |
GenBank mRNA | BC149675.1 | - | 6 - 7 | 739 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
