Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113572.1 | NP_001107044.1 | 1 - 2 | 304 | 82 | |
GenBank mRNA | CO878421.1 | - | 1 - 2 | 195 | 82 | |
DY181718.1 | - | 1 - 2 | 304 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
