Assay Details
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113572.1 | NP_001107044.1 | 3 - 4 | 501 | 137 | |
GenBank mRNA | CO878421.1 | - | 3 - 4 | 392 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
