Assay Details
Gene Symbol: PLA2G2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025324.2 | NP_001020495.1 | 1 - 2 | 64 | 145 | |
GenBank mRNA | AY911335.1 | - | 1 - 2 | 65 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025324.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
