Assay Details
Gene Symbol: OAS1X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178108.2 | NP_835209.1 | 3 - 4 | 688 | 141 | |
GenBank mRNA | AB104656.1 | - | 3 - 4 | 688 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
