Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105001.2 | NP_001098471.1 | 8 | 1753 | 119 | |
GenBank mRNA | BC151514.1 | - | 8 | 1754 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
