Assay Details
Gene Symbol: COX7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175795.3 | NP_786989.3 | 2 - 3 | 295 | 132 | |
GenBank mRNA | BC103174.1 | - | 1 | 223 | 132 | |
EE243932.1 | - | 2 - 3 | 295 | 132 | ||
EH141674.1 | - | 2 - 3 | 235 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
