Assay Details
Gene Symbol: PRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143735.1 | NP_001137207.1 | 1 - 2 | 539 | 108 | |
GenBank mRNA | AY313172.1 | - | 1 - 2 | 360 | 108 | |
FJ176744.1 | - | 1 - 2 | 539 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
