Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 16 - 17 | 2035 | 72 | |
GenBank mRNA | GH316016.1 | - | 2 - 3 | 131 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
