Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 20 - 21 | 2691 | 125 | |
GenBank mRNA | BF075592.1 | - | 3 - 4 | 414 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() |
