Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143857.3 | NP_001137329.3 | 27 - 28 | 3436 | 97 | |
GenBank mRNA | EE354110.1 | - | 2 - 3 | 152 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
