Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY265991.1 | - | 1 - 2 | 720 | 60 | |
BC114027.1 | - | 2 - 3 | 889 | 60 | ||
BC146111.1 | - | 2 - 3 | 889 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
