Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100382.1 | NP_001093852.1 | 1 - 2 | 108 | 59 | |
GenBank mRNA | BC149143.1 | - | 1 - 2 | 108 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
