Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100382.1 | NP_001093852.1 | 9 - 10 | 978 | 92 | |
GenBank mRNA | BC149143.1 | - | 9 - 10 | 978 | 92 | |
BT029847.1 | - | 8 - 9 | 822 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
