Assay Details
Gene Symbol: TRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101221.1 | NP_001094691.1 | 256 | 84 | ||
GenBank mRNA | BC148157.1 | - | 256 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC148157.1, NM_001101221.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
