Assay Details
Gene Symbol: NGFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102478.2 | NP_001095948.1 | 3 - 4 | 690 | 73 | |
GenBank mRNA | BC142255.1 | - | 3 - 4 | 691 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
