Assay Details
Gene Symbol: NCEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123034.1 | NP_001116506.1 | 3 - 4 | 514 | 90 | |
GenBank mRNA | BC115993.1 | - | 3 - 4 | 514 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
