Assay Details
Gene Symbol: NFKBIE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130746.2 | NP_001124218.1 | 1 - 2 | 585 | 101 | |
GenBank mRNA | BC120269.1 | - | 1 - 2 | 586 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
