Assay Details
Gene Symbol: PPP6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130750.1 | NP_001124222.1 | 4 - 5 | 443 | 86 | |
GenBank mRNA | BC112686.1 | - | 4 - 5 | 443 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
