Assay Details
Gene Symbol: COX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076861.1 | NP_001070329.1 | 8 - 9 | 1184 | 88 | |
GenBank mRNA | BC123760.1 | - | 8 - 9 | 1184 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
