Assay Details
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101865.2 | NP_001095335.1 | 2 - 3 | 348 | 91 | |
GenBank mRNA | BC112801.1 | - | 1 - 2 | 145 | 91 | |
BC147963.1 | - | 2 - 3 | 349 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
