Assay Details
Gene Symbol: CCKAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101865.2 | NP_001095335.1 | 5 - 6 | 984 | 108 | |
GenBank mRNA | BC112801.1 | - | 4 - 5 | 781 | 108 | |
BC147963.1 | - | 5 - 6 | 985 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
