Assay Details
Gene Symbol: RCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103221.1 | NP_001096691.1 | 5 - 6 | 959 | 84 | |
GenBank mRNA | BC134456.1 | - | 5 - 6 | 959 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
