Assay Details
Gene Symbol: ALG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103346.1 | NP_001096816.1 | 2 - 3 | 382 | 93 | |
GenBank mRNA | BC142132.1 | - | 2 - 3 | 382 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
