Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163225.1 | NP_001156697.1 | 2 - 3 | 351 | 82 | |
GenBank mRNA | BC103044.1 | - | 2 - 3 | 126 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
