Assay Details
Gene Symbol: VIRMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205426.1 | NP_001192355.1 | 1 - 2 | 82 | 88 | |
GenBank mRNA | BC148879.1 | - | 1 - 2 | 70 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
