Assay Details
Gene Symbol: VIRMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205426.1 | NP_001192355.1 | 22 - 23 | 5154 | 91 | |
GenBank mRNA | BC133316.1 | - | 8 - 9 | 1452 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
