Assay Details
Gene Symbol: RMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205470.1 | NP_001192399.1 | 10 - 11 | 1105 | 103 | |
GenBank mRNA | BC151667.1 | - | 10 - 11 | 1077 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
