Assay Details
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205560.1 | NP_001192489.1 | 2 - 3 | 213 | 104 | |
GenBank mRNA | AY253831.1 | - | 1 - 2 | 77 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
