Assay Details
Gene Symbol: ZKSCAN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205670.1 | NP_001192599.1 | 5 - 6 | 1493 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
