Assay Details
Gene Symbol: CCDC181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205801.1 | NP_001192730.1 | 1 - 2 | 118 | 102 | |
GenBank mRNA | BC109684.1 | - | 2 - 3 | 278 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
