Assay Details
Gene Symbol: ADGRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205846.1 | NP_001192775.1 | 2 - 3 | 325 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001205846.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
