Assay Details
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206080.1 | NP_001193009.1 | 5 - 6 | 1004 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: OPN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206080.1 | NP_001193009.1 | 5 - 6 | 1004 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |