Assay Details
Gene Symbol: PTP4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206124.2 | NP_001193053.1 | 2 - 3 | 756 | 76 | |
NM_001354592.1 | NP_001341521.1 | 2 - 3 | 730 | 76 | ||
GenBank mRNA | BC109916.1 | - | 2 - 3 | 618 | 76 | |
EH374978.1 | - | 1 - 2 | 345 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
