Assay Details
Gene Symbol: CHRNA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192546.1 | NP_001179475.1 | 4 - 5 | 900 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
