Assay Details
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192550.1 | NP_001179479.1 | 32 - 33 | 4676 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NBAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192550.1 | NP_001179479.1 | 32 - 33 | 4676 | 89 |
Set Membership: |
![]() ![]() ![]() ![]() |