Assay Details
Gene Symbol: OFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192637.2 | NP_001179566.2 | 19 - 20 | 2701 | 61 | |
GenBank mRNA | CB172642.1 | - | 4 - 5 | 531 | 61 | |
CB222512.1 | - | 4 - 5 | 495 | 61 | ||
JN193530.1 | - | 20 - 21 | 2790 | 61 | ||
JN193531.1 | - | 20 - 21 | 2883 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
