Assay Details
Gene Symbol: GPR158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206442.1 | NP_001193371.1 | 4 - 5 | 1338 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
