Assay Details
Gene Symbol: AASDHPPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206477.1 | NP_001193406.1 | 2 - 3 | 479 | 129 | |
GenBank mRNA | BC102625.1 | - | 2 - 3 | 448 | 129 | |
DT813625.1 | - | 2 - 3 | 401 | 129 | ||
DT860683.1 | - | 2 - 3 | 480 | 129 | ||
EV716630.1 | - | 4 - 5 | 688 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
