Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206503.1 | NP_001193432.1 | 2 - 3 | 691 | 69 | |
GenBank mRNA | BC149786.1 | - | 2 - 3 | 652 | 69 | |
HQ402594.1 | - | 1 - 2 | 395 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
