Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206503.1 | NP_001193432.1 | 4 - 5 | 1340 | 76 | |
GenBank mRNA | BC149786.1 | - | 4 - 5 | 1301 | 76 | |
HQ402594.1 | - | 3 - 4 | 1044 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
