Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206503.1 | NP_001193432.1 | 6 - 7 | 1724 | 73 | |
GenBank mRNA | BC149786.1 | - | 6 - 7 | 1685 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
