Assay Details
Gene Symbol: RAB11FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192771.1 | NP_001179700.1 | 1 - 2 | 650 | 98 | |
GenBank mRNA | BC151810.1 | - | 1 - 2 | 650 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
